
Baicalein 7-diglucoside

Synonym(s):
Oroxin B, 5,6,7-Trihydroxyflavone 7-diglucoside, Baicalein 7-gentiobioside, Baicalein-7-O-diglucoside
Empirical Formula (Hill Notation):
C27H30O15
CAS Number:
Molecular Weight:
594.52
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing