Skip to Content
Merck
All Photos(1)

Key Documents

91241

Supelco

3-Hydroxy-2-methylbutyric acid, mixture of isomers

analytical standard

Synonym(s):

3-Hydroxy-2-methylbutanoic acid, Nilic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O3
CAS Number:
Molecular Weight:
118.13
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97% (GC)

shelf life

limited shelf life, expiry date on the label

storage temp.

2-8°C

SMILES string

OC(C(C)C(C)O)=O

InChI

1S/C5H10O3/c1-3(4(2)6)5(7)8/h3-4,6H,1-2H3,(H,7,8)

InChI key

VEXDRERIMPLZLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service