Z743564
Petri-Seal™ adhesive sealing film
clear, L × W 12.5 in. × 32.9 in.
All Photos(1)
About This Item
UNSPSC Code:
41122410
NACRES:
NB.35
Recommended Products
sterility
non-sterile
packaging
pack of 1 ea
manufacturer/tradename
Diversified Biotech PSEAL-108C
L × W
12.5 in. × 32.9 in.
color
clear
Related Categories
General description
A sealing tape designed specifically for petri dishes and multiwell plates.
Conforms to the irregular perimeter to seal cover to plate, reduce evaporation, and prevent contamination. Waterproof, chemically resistant, and suitable for continuous use from −40 to +77°C. Heavy 5-mil (0.125 mm) thickness resists abrasion and puncture.
Conforms to the irregular perimeter to seal cover to plate, reduce evaporation, and prevent contamination. Waterproof, chemically resistant, and suitable for continuous use from −40 to +77°C. Heavy 5-mil (0.125 mm) thickness resists abrasion and puncture.
Legal Information
Petri-Seal is a trademark of C.B.S. Scientific Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)