Z741759
Peristaltic pump
4-Channel, AC/DC input 120 V AC, US Plug
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105102
manufacturer/tradename
IBI ACCFL0011
measuring range
resolution: 0.1 rpm
parameter
0.4 rpm speed (45 rpm)
Looking for similar products? Visit Product Comparison Guide
General description
IBI offers a high quality 4-channel peristaltic pump for use with the IBI flow cell systems. The 4-channel pump can accommodate one each of the 3-channel flow cells or up to four each of the convertible flow cells.
The pump has a unique planetary gear drive system with 8 rollers that provides the most gentle fluid delivery of any multi-channel peristaltic pump which is ideal for use with the IBI flow cell systems. The pump utilizes stoppered tubing that accommodates the cassette design of the pump. IBI flow cell assemblies come complete with the necessary pump tubing already in place. (Except cell ONLY models).
The pump has a unique planetary gear drive system with 8 rollers that provides the most gentle fluid delivery of any multi-channel peristaltic pump which is ideal for use with the IBI flow cell systems. The pump utilizes stoppered tubing that accommodates the cassette design of the pump. IBI flow cell assemblies come complete with the necessary pump tubing already in place. (Except cell ONLY models).
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service