SCYT-91K-PX14
MILLIPLEX® Ovine Cytokine/Chemokine Panel 1 Premixed 14-plex - Immunology Multiplex Assay
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161503
NACRES:
NA.47
species reactivity
sheep
manufacturer/tradename
Milliplex®
technique(s)
multiplexing: suitable
detection method
fluorometric (Luminex® xMAP®)
storage temp.
2-8°C
General description
Inflammation and immune cell response are key to maintaining homeostasis. Inflammation plays a central role in the etiology of many immunologically related diseases, such as infectious disease, osteoarthritis and respiratory disease, as well as neurologic, metabolic and cardiovascular disease and even cancer. However, until now the ability to measure levels of cytokines and chemokines in sheep samples has been limited primarily to ELISAs. Researchers now have the ability to measure multiple inflammation biomarkers simultaneously using our MILLIPLEX® Ovine Cytokine/Chemokine Panel 1, based on the Luminex xMAP technology.MILLIPLEX® Ovine Cytokine/Chemokine Panel 1 may be used for the simultaneous quantification of any or all of the following analytes in serum, plasma, and tissue culture supernatant samples: IFNγ, IL-1α, IL-1β, IL-4, IL-6, IL-8 (CXCL8), IL-10, IL-17A, IL-36RA (IL-1F5), IP-10 (CXCL10), MIP-1α (CCL3), MIP-1β (CCL4), TNFα, VEGF-A. Note: The protocol recommends a 1:2 dilution of serum and plasma samples, except when assaying IL-8 (CXCL8) in serum samples for which an up to 1:8 dilution of sample may be required.MILLIPLEX® offers the broadest selection of analytes across a wide range of disease states and species. Once the analytes of interest have been identified, you can rely on the quality that we build into each kit to produce results you can trust. In addition to the assay characteristics listed in the protocol, other performance criteria evaluated during the verification process include: cross-reactivity, dilution linearity, kit stability, and sample behavior (e.g. detectability and stability). In addition each panel and kit meets stringent manufacturing criteria to ensure batch-to-batch reproducibility.Each MILLIPLEX® panel and kit includes:Quality controls (QCs) to qualify assay performanceComparison of standard (calibrator) and QC lots to a reference lot to ensure lot-to-lot consistencyOptimized serum matrix to mimic native analyte environment in serum and plasma samplesDetection antibody cocktails designed to yield consistent analyte profiles within panelThe MILLIPLEX® Ovine Cytokine/Chemokine Panel 1 thus enables you to focus on the therapeutic potential of cytokines and the modulation of cytokine expression.
Specificity
Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.
Application
The Ovine Cytokine/Chemokine bead-based multiplex cytokine panel, using the Luminex® xMAP technology, enables the simultaneous analysis of 14 cytokine and chemokine biomarkers in ovine serum, plasma and cell culture samples.This is an overnight incubation assay.The protocol recommends a 1:2 dilution of serum and plasma samples, except when assaying IL-8 (CXCL8) in serum samples for which an up to 1:8 dilution of sample may be required.This kit may be used for the analysis of any or all of the Ovine Cytokine/Chemokine Panel 1 analytes in ovine tissue/cell lysate and culture supernatant samples and serum or plasma samples.Analytes available:IFN-γ;IL-1α;IL-1β;IL-4;IL-6;IL-10;IL-8;IL-17A;MIP-1α;IL-36RA;IP-10;MIP-1β;TNFα;VEGF-A
Features and Benefits
Configuration: Premixed
Packaging
96-well plate
Storage and Stability
Recommended storage for kit components is 2 - 8°C.
Legal Information
Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2
Target Organs
Respiratory Tract
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service