Skip to Content
Merck
All Photos(3)

Documents

MIAABG100

Millipore

Microfil® funnel with S-Pak®

sample volume 100 mL, mixed cellulose esters (MCE), pore size 0.8 μm, filter diam. 47 mm, black filter, gridded filter, To detect yeast and mold

Synonym(s):

Membrane filtration system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

material

black filter
gridded filter
mixed cellulose esters (MCE)
polypropylene

Agency

ISO 7704

sterility

sterile; ethylene oxide treated

feature

easily wetted with water
hydrophilic

manufacturer/tradename

Microfil® with S-Pak®

parameter

100 mL sample volume

filter diam.

47 mm

matrix

MF-Millipore

pore size

0.8 μm pore size

input

sample type beverage(s)
sample type food(s)
sample(s) (environmental)

application(s)

bioburden testing
clinical
cosmetics
environmental
environmental
food and beverages
industrial qc

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

The Microfil filtration system is a simple and reliable solution for routine microbiological testing. These ready-to-use, 100 mL Microfil funnels that comes with the 0.8 μm pore size, black gridded S pak filter system eliminate the need for washing and sterilizing after each test thus making the equipment handling easier and efficient to retain and recover microorganisms like yeast and molds. The push-fit design of the Microfil funnels seals tightly to manifold supports without the need for clamps or O-rings. Unlike with conventional filter holders, incomplete sealing at the membrane/funnel interface is eliminated resulting in consistent recovery of organisms.

Application

These filters find application in Clinical, Cosmetics, Environmental, Food and Beverage, Industrial QC and Pharmaceutical industry. Microbial analysis of beer, bottled water, cider, sport drink and wine by membrane filtration method

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany
S-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service