Skip to Content
Merck
All Photos(1)

Key Documents

8.41690

Sigma-Aldrich

Chrysene

for synthesis

Synonym(s):

Chrysene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12
CAS Number:
Molecular Weight:
228.29
MDL number:
UNSPSC Code:
12352127
EC Index Number:
205-923-4
NACRES:
NA.22

Quality Level

Assay

≥95.0% (HPLC)

form

crystals

bp

448 °C

mp

256-258 °C

solubility

<0.0001 g/L

density

1.274 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

c21c(c4c(cc2)cccc4)ccc3c1cccc3

InChI

1S/C18H12/c1-3-7-15-13(5-1)9-11-18-16-8-4-2-6-14(16)10-12-17(15)18/h1-12H

InChI key

WDECIBYCCFPHNR-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 95.0 % (a/a)
Melting range (lower value): ≥ 252 °C
Melting range (upper value): ≤ 256 °C
Identity (IR): passes test

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service