Skip to Content
Merck
All Photos(1)

Documents

494127-M

Millipore

Ghrelin, Rat, Mouse, Synthetic

The 28-amino acid n-octanoylated (at ser3) natural peptidyl ligand for the growth hormone secretagogue receptor 1a (GHSR1a).

Synonym(s):

Ghrelin, Rat, Mouse, Synthetic, GS-S(n-octanoyl)-FLSPEHQKAQQRKESKKPPAKLQPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C147H245N45O42
Molecular Weight:
3314.79
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 1 mg/mL

storage temp.

2-8°C

General description

The 28-amino acid n-octanoylated (at ser3) natural peptidyl ligand for the growth hormone secretagogue receptor 1a (GHSR1a). Ghrelin is secreted predominantly in the stomach and stimulates growth hormone release from the pituitary gland. Rat and mouse ghrelin differ from that of human (Cat. No. 494126) by only two amino acids.

Warning

Toxicity: Standard Handling (A)

Sequence

H-Gly-Ser-Ser(Octanoyl)-Phe-Leu-Ser-Pro-Glu-His-Gln-Lys-Ala-Gln-Gln-Arg-Lys-Glu-Ser-Lys-Lys-Pro-Pro-Ala-Lys-Leu-Gln-Pro-Arg-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Ukkola, O. 2009. Curr. Protein Pept. Sci.10, 2.
Kojima, M., et al. 1999. Nature402, 656.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service