Merck
  • Deposition of a photosensitive complex within a lecithin bilayer lipid membrane.
Deposition of a photosensitive complex within a lecithin bilayer lipid membrane.
Journal of photochemistry and photobiology. B, Biology (1989-06-01)
Z Salamon, H T Tien
ABSTRACT
In this paper the phenomenon of a photosensitive ion complex of Brilliant Yellow and ferric ions formation in the electrolyte phase and its subsequent deposition within a bilayer lipid membrane (BLM) is described. Deposition of light sensitive complex into the BLM considerably increases its mechanical stability and drastically changes its electrochemical and photoelectrical properties as well.
Sigma-Aldrich
Brilliant Yellow, Dye content ≥50 %