MilliporeSigma
  • beta-Adrenoceptor regulation in rat heart, lung and skin after chronic treatment with (--)-tertatolol or (--)-propranolol.
beta-Adrenoceptor regulation in rat heart, lung and skin after chronic treatment with (--)-tertatolol or (--)-propranolol.
Journal of autonomic pharmacology (1995-12-01)
Y Y Tan, R J Summers
ABSTRACT
1. The effect of long-term treatment with the beta-adrenoceptor antagonists (--)-tertatolol and (--)-propranolol was studied. Sprague-Dawley rats were treated with either (--)-tertatolol (50 micrograms kg-1 hr-1), (--)-propranolol (250 micrograms kg-1 hr-1) or vehicle (1 mM HCl) for 14 days with osmotic minipumps implanted subcutaneously. 2. The mean daily systolic blood pressure and heart rate of rats treated with either (--)-tertatolol (108 +/- 1 mmHg/330 +/- 3 bpm) or (--)-propranolol (103 +/- 1 mmHg/330 +/- 2 bpm) were lower than in the control (126 +/- 1 mmHg/405 +/- 3 bpm, P < 0.001, n = 8-10) indicating the effectiveness of drug delivery. 3. Autoradiographic studies in areas of heart, lung and skin showed that beta-adrenoceptor populations were not significantly affected by the drug treatment (all regions P > 0.05). Nevertheless, the receptor population in the homogenates of (--)-tertatolol treated lung were halved (194 +/- 28 fmol mg protein-1 compared with a control value of 388 +/- 54 fmol mg protein-1, P < 0.01, n = 6). 4. In the presence of CGP 20712A, the left atrial inotropic and right atrial chronotropic responsiveness to (--)-isoprenaline were hypersensitive in both (--)-tertatolol and (--)-propranolol-treated groups (P < 0.005, ANCOVA). 5. (--)-Propranolol treated left ventricular free wall had lower basal [3H]-forskolin binding to adenylate cyclase (14.45 +/- 1.20 fmol mg protein-1 compared with a control value of 18.91 +/- 0.78 fmol mg protein-1, P = 0.01, n = 6). (--)-Tertatolol treatment had no effect on the basal binding. In the presence of the G-protein activators NaF and Gpp(NH)p, the enhancement of [3H]-forskolin binding did not differ between control and the drug treated groups. 6. Chronic (--)-tertatolol or (--)-propranolol treatment therefore did not produce an increase in receptors in heart, lung or skin but the beta-adrenoceptor-mediated responses were enhanced. In addition, [3H]-forskolin binding did not increase suggesting that the hypersensitivity was not due to changes in the number of receptors or adenylate cyclase. Hypersensitivity following beta-adrenoreceptor antagonist administration may therefore involve enhanced coupling of receptors to G-proteins.
Sigma-Aldrich
(±)-Propranolol hydrochloride, ≥99% (TLC), powder