
2′,7′-Dichlorodihydrofluorescein diacetate

Synonym(s):
2′,7′-Dichlorofluorescin diacetate, 2′,7′-Dichlorodihydrofluorescein diacetate
Empirical Formula (Hill Notation):
C24H16Cl2O7
CAS Number:
Molecular Weight:
487.29
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing