TMO3410-08
Nalgene®PC Biotainer® Bottles and Carboys
clear polycarbonate, 10000 mL capacity, case of 2 ea
Synonym(s):
PETG bottle, bottle with handle, square bottle
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Recommended Products
material
clear polycarbonate
sterility
sterile
packaging
case of 2 ea
manufacturer/tradename
Nalgene 3410-08
capacity
10000 mL
10000 mL capacity
Related Categories
General description
Eliminate the cost of cleaning, sterilizing and associated validations compared to bottles designed for reuse with Thermo Scientific Nalgene PETG Biotainer Bottles, ideal for prolonged storage of pH sensitive materials such as culture media. Certified sterile and ready-to-use, all Biotainer bottles are gamma irradiated to a Sterility Assurance Level (SAL) of 10–6.
Legal Information
Biotainer is a registered trademark of In Vitro Scientific Products
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)