Skip to Content
Merck
All Photos(1)

Key Documents

SRP0699

Sigma-Aldrich

STK9 Protein

recombinant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

recombinant

expressed in baculovirus infected Sf9 cells

Assay

70-95% (SDS-PAGE)

form

frozen liquid

mol wt

~84 kDa

concentration

0.1 μg/μL

accession no.

NM_003159

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... STK9(6792)

Application

Useful for Western blot.

Biochem/physiol Actions

Recombinant human STK9 (CDKL5) (1-498) was expressed by Baculovirus in Sf9 insect cells using an N-terminal GST tag. STK9 or CDKL5 is a member of Ser/Thr protein kinase family which has protein kinase activity and is associated with X-linked infantile spasm syndrome (ISSX), also known as X-linked West syndrome, and Rett syndrome (RTT). STK9 can control nuclear speckle morphology by regulating the phosphorylation state of splicing regulatory proteins, and may be involved indirectly in pre-mRNA processing, by controlling splicing factor dynamics (1). The mutations in the STK9 is a major cause of infantile spasms and early epileptic seizures in female patients, and of a later intractable seizure disorder (2).

Reconstitution

Formulated in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.25 mM DTT, 0.1 mM PMSF, with 25% glycerol.

Storage and Stability

Store in aliquots at -70°C. Avoid freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service