MTCT8842
MTC™ Bio DuoClick™ Two Position Screw-cap Culture Tube
capacity 14 mL, polystyrene, sterile, pack of 500 ea (10 x racks 50EA)
Synonym(s):
culture test tube, plastic culture tube, plastic tube, sample mailing tube
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121706
NACRES:
NB.15
Recommended Products
material
polystyrene
round
screw cap
sterility
sterile
feature
graduation
packaging
pack of 500 ea (10 x racks 50EA)
manufacturer/tradename
MTC™ Bio T8842
parameter
1400 × g speed
capacity
14 mL
diam. × L
17 mm × 100 mm
suitability
suitable for (cell culture)
Looking for similar products? Visit Product Comparison Guide
General description
Culture Tube, 14mL, 17 x 100mm, PS, w/ attached 2-position screw-cap, printed graduations, sterile, 10 foam racks of 50 tubes, 500/case
MTC™ Bio’s DuoClick™ tubes feature a unique, patented screw closure system. DuoClick™ tubes completely eliminate the excess force, splashing and general frustration of traditional snap-cap culture tubes. The advanced cap design includes two distinct positions for aerobic and anaerobic culturing. Rotate the cap to the first click to secure the cap in the aerobic position or, rotate past the second click for a leak-proof, airtight seal for anaerobic culturing.
DuoClick™ tubes are available e-beam sterilized, with screw caps attached, and in ready-to-use rack packaging.
DuoClick™ tubes are available e-beam sterilized, with screw caps attached, and in ready-to-use rack packaging.
Application
aerobic andanaerobic culturing
Features and Benefits
- New patent pending screw-cap!
- Aerobic culturing at first click
- Anaerobic after second click
- Printed graduations
- Flat, markable screw-cap
Legal Information
DuoClick is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service