Skip to Content
Merck
All Photos(1)

Documents

MSST0056

Sigma-Aldrich

SILuLite APOB48, Apolipoprotein B-48 human

recombinant, expressed in HEK 293 cells, MS Protein Standard

Synonym(s):

APOB48, Apo B-48, Mass spectrometry standard, Apo B-48, Mass spectrometry standard, Apolipoprotein B-48

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
NACRES:
NA.32

recombinant

expressed in HEK 293 cells

Quality Level

tag

FLAG® tagged
His tagged

Assay

≥95% (SDS-PAGE)

form

liquid

suitability

suitable for mass spectrometry (internal calibrator)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... APOB48(338)

General description

Apolipoprotein B 48 (APOB48) is a key subunit of APOB. APOB is encoded by the gene mapped to human chromosome 2p24.1.
SILuLite APOB48 is a recombinant human protein expressed in human 293 cells. It consists of 2172 amino acids, with a calculated molecular mass of 243.2 kDa (including C-terminal polyhistidine and flag tags). SILuLite APOB48 is an analytical standard designed to be used as starting material for preparation of calibrators and controls in LC-MS applications.

Biochem/physiol Actions

Apolipoprotein B 48 acts as a ligand for low-density lipoprotein (LDL)-receptor. Elevated levels of APOB is associated with the development of elevated LDL levels and other lipid metabolic disorders. In addition, overexpression of the gene increases the risk of susceptibility to coronary heart disease.
Apolipoprotein B-48 (Apo B-48) is known to be the only specific marker of intestinal chylomicron (CM) particles. The amino acid sequence of apoB-48 represents 48% of the initial sequence of apolipoprotein B-100 (Apo B-100). Apo B-48 is synthesized only by the intestine in humans, while Apo B-100 is synthesized primarily by the liver. Therefore, Apo B-48 is a most appropriate biomarker for cardiovascular and nutritional investigation of postprandial CM metabolism.In a recent study, Apo B-48 was discovered as a novel biomarker in amyotrophic lateral sclerosis patient fibroblasts.

Sequence

EEEMLENVSLVCPKDATRFKHLRKYTYNYEAESSSGVPGTADSRSATRINCKVELEVPQLCSFILKTSQCTLKEVYGFNPEGKALLKKTKNSEEFAAAMSRYELKLAIPEGKQVFLYPEKDEPTYILNIKRGIISALLVPPETEEAKQVLFLDTVYGNCSTHFTVKTRKGNVATEISTERDLGQCDRFKPIRTGISPLALIKGMTRPLSTLISSSQSCQYTLDAKRKHVAEAICKEQHLFLPFSYKNKYGMVAQVTQTLKLEDTPKINSRFFGEGTKKMGLAFESTKSTSPPKQAEAVLKTLQELKKLTISEQNIQRANLFNKLVTELRGLSDEAVTSLLPQLIEVSSPITLQALVQCGQPQCSTHILQWLKRVHANPLLIDVVTYLVALIPEPSAQQLREIFNMARDQRSRATLYALSHAVNNYHKTNPTGTQELLDIANYLMEQIQDDCTGDEDYTYLILRVIGNMGQTMEQLTPELKSSILKCVQSTKPSLMIQKAAIQALRKMEPKDKDQEVLLQTFLDDASPGDKRLAAYLMLMRSPSQADINKIVQILPWEQNEQVKNFVASHIANILNSEELDIQDLKKLVKEALKESQLPTVMDFRKFSRNYQLYKSVSLPSLDPASAKIEGNLIFDPNNYLPKESMLKTTLTAFGFASADLIEIGLEGKGFEPTLEALFGKQGFFPDSVNKALYWVNGQVPDGVSKVLVDHFGYTKDDKHEQDMVNGIMLSVEKLIKDLKSKEVPEARAYLRILGEELGFASLHDLQLLGKLLLMGARTLQGIPQMIGEVIRKGSKNDFFLHYIFMENAFELPTGAGLQLQISSSGVIAPGAKAGVKLEVANMQAELVAKPSVSVEFVTNMGIIIPDFARSGVQMNTNFFHESGLEAHVALKAGKLKFIIPSPKRPVKLLSGGNTLHLVSTTKTEVIPPLIENRQSWSVCKQVFPGLNYCTSGAYSNASSTDSASYYPLTGDTRLELELRPTGEIEQYSVSATYELQREDRALVDTLKFVTQAEGAKQTEATMTFKYNRQSMTLSSEVQIPDFDVDLGTILRVNDESTEGKTSYRLTLDIQNKKITEVALMGHLSCDTKEERKIKGVISIPRLQAEARSEILAHWSPAKLLLQMDSSATAYGSTVSKRVAWHYDEEKIEFEWNTGTNVDTKKMTSNFPVDLSDYPKSLHMYANRLLDHRVPQTDMTFRHVGSKLIVAMSSWLQKASGSLPYTQTLQDHLNSLKEFNLQNMGLPDFHIPENLFLKSDGRVKYTLNKNSLKIEIPLPFGGKSSRDLKMLETVRTPALHFKSVGFHLPSREFQVPTFTIPKLYQLQVPLLGVLDLSTNVYSNLYNWSASYSGGNTSTDHFSLRARYHMKADSVVDLLSYNVQGSGETTYDHKNTFTLSYDGSLRHKFLDSNIKFSHVEKLGNNPVSKGLLIFDASSSWGPQMSASVHLDSKKKQHLFVKEVKIDGQFRVSSFYAKGTYGLSCQRDPNTGRLNGESNLRFNSSYLQGTNQITGRYEDGTLSLTSTSDLQSGIIKNTASLKYENYELTLKSDTNGKYKNFATSNKMDMTFSKQNALLRSEYQADYESLRFFSLLSGSLNSHGLELNADILGTDKINSGAHKATLRIGQDGISTSATTNLKCSLLVLENELNAELGLSGASMKLTTNGRFREHNAKFSLDGKAALTELSLGSAYQAMILGVDSKNIFNFKVSQEGLKLSNDMMGSYAEMKFDHTNSLNIAGLSLDFSSKLDNIYSSDKFYKQTVNLQLQPYSLVTTLNSDLKYNALDLTNNGKLRLEPLKLHVAGNLKGAYQNNEIKHIYAISSAALSASYKADTVAKVQGVEFSHRLNTDIAGLASAIDMSTNYNSDSLHFSNVFRSVMAPFTMTIDAHTNGNGKLALWGEHTGQLYSKFLLKAEPLAFTFSHDYKGSTSHHLVSRKSISAALEHKVSALLTPAEQTGTWKLKTQFNNNEYSQDLDAYNTKDKIGVELTGRTLADLTLLDSPIKVPLLLSEPINIIDALEMRDAVEKPQEFTIVAFVKYDKNQDVHSINLPFFETLQEYFERNRQTIIVVLENVQRNLKHINIDQFVRKYRAALGKLPQQANDYLNSFNWERQVSHAKEKLTALTKKYRITENDIQIALDDAKINFNEKLSQLQTYMIDYKDDDDKGHHHHHHHHGGQ

Physical form

Supplied as a liquid containing phosphate buffered saline.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association between polymorphisms in the APOB gene and hyperlipidemia in the Chinese Yugur population
Gu Q L, et al.
Brazilian journal of medical and biological research, 50(11) (2017)
Forty-three loci associated with plasma lipoprotein size, concentration, and cholesterol content in genome-wide analysis.
Chasman DI, et al.
PLoS Genetics, 5(11), e1000730-e1000730 (2009)
Meta-analysis of four new genome scans for lipid parameters and analysis of positional candidates in positive linkage regions.
Heijmans BT
European Journal of Human Genetics, 13(10), 1143-1153 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service