CLS3006
Corning® single use spatulas
V-scoop/spoon
Synonym(s):
Disposable spatulas
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122403
NACRES:
NB.14
Recommended Products
sterility
sterile
manufacturer/tradename
Corning 3006
L
250 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Scoop dimensions:
Maximum scoop width - 0.13 inches (3.302 mm)
scoop can hold < 0.5 mL
Maximum scoop width - 0.13 inches (3.302 mm)
scoop can hold < 0.5 mL
Corning single use spatulas have been designed to save time and provide contamination-free samples. Eliminates need to recycle and sterilize reusable spatulas.
Each spatula is individually packaged, RNase/DNase free, non-pyrogenic and antistatic.
Each spatula is individually packaged, RNase/DNase free, non-pyrogenic and antistatic.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service