1.28204
Sodium acetate trihydrate
EMPROVE® API, Ph. Eur., BP, JP, USP, FCC, E 262
Pharma Manufacturing
Synonym(s):
Acetic acid sodium salt trihydrate
About This Item
Recommended Products
Agency
BP
JP
Ph. Eur.
USP
Quality Level
reg. compliance
FCC
product line
EMPROVE® API
form
crystalline
autoignition temp.
1112 °F
application(s)
pharmaceutical
storage temp.
2-25°C
SMILES string
O.O.O.[Na+].CC([O-])=O
InChI
1S/C2H4O2.Na.3H2O/c1-2(3)4;;;;/h1H3,(H,3,4);;3*1H2/q;+1;;;/p-1
InChI key
AYRVGWHSXIMRAB-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
Application
Other Notes
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
What is the Emprove® Program?
The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:
Available by subscription or for a fee
Free of charge - Sign in to download
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service