Key Documents
Z742268
DURAN® TILT GL 56 self-adhesive identification label for screw cap
diam. 40 mm, white polyester, pack of 60 ea (6 labels per sheet)
Synonym(s):
294015604
About This Item
UNSPSC Code:
55121611
NACRES:
NB.17
material
white polyester
packaging
pack of 60 ea (6 labels per sheet)
manufacturer/tradename
DURAN® 294015604
diam.
40 mm
General description
Careful labeling is very important to prevent mix ups and mistakes. The GL 56 self-adhesive cap labels can be used to clearly indicate the separate bottles of media for each cell line, preventing possible cross-contamination.
Features and Benefits
  • Manufactured from polyester
  • Can be used to clearly indicate the separate bottles of media for each cell line, preventing possible cross-contamination
  • High temperature performance range makes them ideal for use in fridges, freezers, autoclaves, incubators, and water baths
  • The labels have a high adhesive strength they are peelable and easy to remove which safes time for cleaning
Legal Information
DURAN is a registered trademark of DWK Life Sciences
accessory
Choose from one of the most recent versions:
Certificates of Analysis (COA)