Skip to Content
Merck
All Photos(1)

Key Documents

Z177822

Scienceware® polypropylene trays

L × W 48 cm × 27 cm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.51

material

polypropylene container

manufacturer/tradename

Bel-Art

L × W

48 cm × 27 cm

capacity

13 L (3.4 gal)

Looking for similar products? Visit Product Comparison Guide

General description

Lightweight molded polypropylene trays have rounded corners and smooth, tapered sides for nesting.

  • Chemical resistant and corrosion proof
  • Ideal for sterilizing instruments, bottles containing agar, deionized water, saline and other reagents, and for safely cleaning glassware in sinks
  • Steam autoclavable at 121°C (250°F)
  • Cover not included

Legal Information

Scienceware is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service