Key Documents
CAD4850
Stainless steel 304 syringe needle
gauge 16, L 4 in.
About This Item
UNSPSC Code:
42142600
NACRES:
NB.03
description
Standard Hub with 304 Stainless Steel Tubing
packaging
pack of 12 ea
manufacturer/tradename
Cadence Science 4850
L
4 in.
gauge
16
General description
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
Choose from one of the most recent versions:
Certificates of Analysis (COA)