Merck
  • Photochemical fishing approaches for identifying target proteins and elucidating the structure of a ligand-binding region using carbene-generating photoreactive probes.
Photochemical fishing approaches for identifying target proteins and elucidating the structure of a ligand-binding region using carbene-generating photoreactive probes.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2006-03-04)
Yutaka Sadakane, Yasumaru Hatanaka
ABSTRACT
Photoaffinity labeling enables the direct probing of a target protein through a covalent bond between a ligand and its binding protein, and even a complex formed by weak interactions can be isolated by the method. The photochemical fishing approach accelerates the throughput, isolating crosslinked complexes and analyzing the structure of the ligand binding site within the protein. We used carbene-generating phenyldiazirine for this approach because practical examinations had shown that the phenyldiazirine functioned as the powerful barb on the hook. Improving the synthetic pathways of the photoprobes and using chemoselective-integrated photoreactive units makes possible the easy and rapid preparation of carbene-generating photoreactive probes including the derivatives in peptides, proteins, DNAs, and carbohydrates. This review also shows several recent impacts of photoaffinity labeling, including the in vivo preparation of photoreactive proteins in living cells.
Sigma-Aldrich
Azidobenzene solution, ~0.5 M in 2-methyltetrahydrofuran, ≥95.0% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Azidobenzene solution, ~0.5 M in tert-butyl methyl ether