Skip to Content
Merck
All Photos(1)

Key Documents

507911

Supelco

Model 24 Single-Stage Regulator

for use with CGA-160 for SCOTTY 14 cylinder (This item does not include the syringe adapter).

Synonym(s):

Model 24 Regulator w/guage, CGA-160, SCOTTY® 14

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500
Pricing and availability is not currently available.

description

single-stage

manufacturer/tradename

Scott Gas

compatibility

for use with CGA-160 for SCOTTY 14 cylinder (This item does not include the syringe adapter).

Looking for similar products? Visit Product Comparison Guide

General description

Designed for use with SCOTTY® 14 cylinders, this compact, lightweight, single-stage regulator has a CGA 160 inlet and is recommended for use with non-corrosive gases.

Features and Benefits

  • Model 24 Single-Stage Regulator is designed for non-corrosive service.
  • Gauge displays remaining cylinder pressure.
  • Regulator adjusts delivery pressure.
  • Brass body with acetal resin bonnet/Viton diaphragm.
  • Tamper-resistant, locking control knob.
  • Inlet connection ¼" AN flare (CGA-165 or CGA-160).
  • Maximum inlet pressure 300psig.
  • Delivery pressure range 1-10 psig (0.1-0.7 bar) can be preset.
  • Miniature size.

Caution

Cannot be used with Oxygen

Legal Information

SCOTTY is a registered trademark of Air Liquide

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Compressed Gas Association
Handbook of Compressed Gases, 48-48 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service