Skip to Content
Merck
All Photos(1)

Key Documents

AXYHT1000CBKHTRS

Axygen CO-RE style liquid level sensing filter tip

capacity 1000 μL, sterile

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.13
volume:
1000 μL
sterility:
γ-irradiated
sterile
feature:
liquid level sensing
packaging:
case of 1536 tips racked
manufacturer/tradename:
Corning HT1000CBKHTRS

sterility

γ-irradiated
sterile

feature

liquid level sensing

packaging

case of 1536 tips racked

manufacturer/tradename

Corning HT1000CBKHTRS

capacity

1000 μL

volume

1000 μL

color

clear tip

Looking for similar products? Visit Product Comparison Guide

Related Categories


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service