Skip to Content
Merck
All Photos(1)

Key Documents

ARK2140

SAFC

Squalene

redistilled, from animal

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H50
CAS Number:
Molecular Weight:
410.72
MDL number:
UNSPSC Code:
41116011

biological source

animal

Quality Level

storage temp.

2-8°C

SMILES string

C(C\C=C(\CCC=C(C)C)/C)\C(=C\CC\C=C(\CC\C=C(\CCC=C(C)C)/C)/C)\C

InChI

1S/C30H50/c1-25(2)15-11-19-29(7)23-13-21-27(5)17-9-10-18-28(6)22-14-24-30(8)20-12-16-26(3)4/h15-18,23-24H,9-14,19-22H2,1-8H3/b27-17+,28-18+,29-23+,30-24+

InChI key

YYGNTYWPHWGJRM-AAJYLUCBSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service