Skip to Content
Merck
All Photos(1)

Key Documents

878126C

Avanti

C18:1 Lyso PAF

Avanti Research - A Croda Brand 878126C

Synonym(s):

1-O-(9Z)octadecenyl-2-hydroxy-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H54NO6P
CAS Number:
Molecular Weight:
507.68
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mL (878126C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 878126C

concentration

5 mg/mL (878126C-5mg)

lipid type

bioactive lipids
phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COCCCCCCCC/C=C\CCCCCCCC

InChI key

XWYSLMAMRKYUFH-HTOVTZSWSA-N

Application

C18:1 Lyso PAF or 1-O-(9Z)octadecenyl-2-hydroxy-sn-glycero-3-phosphocholine may be used to study the substrate specificity of P.putida KT2440 lyPlsase.

Packaging

5 mL Clear Glass Sealed Ampule (878126C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advances in Clinical Chemistry (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service