Skip to Content
Merck
All Photos(1)

Key Documents

932795

Sigma-Aldrich

Tweezers

Style 2AB, ESD Rubber Coated

Synonym(s):

Tweezer, Grippers, Tweezers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NB.35
Pricing and availability is not currently available.

General description

EMS techware

Application

Safe ergonomic cushion grips (resistivity 10E9 Ohm) for handling ESD sensitive components or small static items. Soft ESD safe cushion grips (resistively 10E8 ohm), these tweezers are ergonomically designed. OAL: 120 mm. Material: Anti-Acid/Anti-MagSS.
Tips: Flat, round, curved, superior finish. Soft ESD safe cushion grips (resistively 10E8 ohm), these tweezers are ergonomically designed.
OAL: 120 mm. Material: Anti-Acid/Anti-MagSS. Tips: Flat, round, curved, superior finish.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service