Skip to Content
Merck
All Photos(1)

Key Documents

45999

Supelco

2-Chloro-4-nitrotoluene

analytical standard, for environmental analysis

Synonym(s):

2-Chloro-1-methyl-4-nitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)Cl
CAS Number:
Molecular Weight:
171.58
Beilstein:
1817924
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Pricing and availability is not currently available.

grade

analytical standard, for environmental analysis
analytical standard

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

60-64 °C (lit.)

application(s)

environmental

SMILES string

Cc1ccc(cc1Cl)[N+]([O-])=O

InChI

1S/C7H6ClNO2/c1-5-2-3-6(9(10)11)4-7(5)8/h2-4H,1H3

InChI key

LLYXJBROWQDVMI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Riedel-de Haën is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service