Merck
  • Large-scale proteomic analysis of tyrosine-phosphorylation induced by T-cell receptor or B-cell receptor activation reveals new signaling pathways.
Large-scale proteomic analysis of tyrosine-phosphorylation induced by T-cell receptor or B-cell receptor activation reveals new signaling pathways.
Proteomics (2009-07-18)
Masaki Matsumoto, Koji Oyamada, Hidehisa Takahashi, Takamichi Sato, Shigetsugu Hatakeyama, Keiichi I Nakayama
ABSTRACT
Activation of the T-cell receptor (TCR) and that of the B-cell receptor (BCR) elicits tyrosine-phosphorylation of proteins that belongs to similar functional categories, but result in distinct cellular responses. Large-scale analyses providing an overview of the signaling pathways downstream of TCR or BCR have not been described, so it has been unclear what components of these pathways are shared and which are specific. We have now performed a systematic analysis and provide a comprehensive list of tyrosine-phosphorylated proteins (PY proteome) with quantitative data on their abundance in T cell, B cell, and nonlymphoid cell lines. Our results led to the identification of novel tyrosine-phosphorylated proteins and signaling pathways not previously implicated in immunoreceptor signal transduction, such as clathrin, zonula occludens 2, eukaryotic translation initiation factor 3, and RhoH, suggesting that TCR or BCR signaling may be linked to downstream processes such as endocytosis, cell adhesion, and translation. Thus comparative and quantitative studies of tyrosine-phosphorylation have the potential to expand knowledge of signaling networks and to promote understanding of signal transduction at the system level.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-ZAP-70 Antibody, clone 1E7.2, clone 1E7.2, Upstate®, from mouse
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Lck Antibody, clone 3A5, clone 3A5, Upstate®, from mouse