
9-Ethylguanine

Synonym(s):
2-Amino-9-ethyl-6-hydroxypurine, 6-Amino-9-ethyl-2-hydroxypurine, 9-EtG
Empirical Formula (Hill Notation):
C7H9N5O
CAS Number:
Molecular Weight:
179.18
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1