1724441
USP
Ziprasidone Related Compound C
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
5,5′-Bis[2-[4-(1,2-benzisothiazol-3-yl)-1-piperazinyl]ethyl]-6,6′-dichloro-1,1′,3,3′-tetrahydro-3-hydroxy-[3,3′-bi-2H-indole]-2,2′-dione, 5,5′-Bis{2-[4-(benzo[d]isothiazol-3-yl)piperazin-1-yl]ethyl}-6,6′-dichloro-3-hydroxy-3,3′-biindoline-2,2′-dione
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
ziprasidone
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
InChI
1S/C42H40Cl2N8O3S2/c43-31-23-33-29(21-25(31)9-11-49-13-17-51(18-14-49)38-27-5-1-3-7-35(27)56-47-38)37(40(53)45-33)42(55)30-22-26(32(44)24-34(30)46-41(42)54)10-12-50-15-19-52(20-16-50)39-28-6-2-4-8-36(28)57-48-39/h1-8,21-24,37,55H,9-20H2,(H,45,53)(H,46,54)
InChI key
JXXKGHVYXULCTC-UHFFFAOYSA-N
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Analysis Note
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service