PHL85956
Cannabigerol
phyproof® Reference Substance
Synonym(s):
CBG
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C21H32O2
CAS Number:
Molecular Weight:
316.48
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥95% (HPLC)
manufacturer/tradename
PhytoLab
storage temp.
2-8°C
SMILES string
Oc1c(c(cc(c1)CCCCC)O)C\C=C(\CCC=C(C)C)/C
InChI
1S/C21H32O2/c1-5-6-7-11-18-14-20(22)19(21(23)15-18)13-12-17(4)10-8-9-16(2)3/h9,12,14-15,22-23H,5-8,10-11,13H2,1-4H3/b17-12+
InChI key
QXACEHWTBCFNSA-SFQUDFHCSA-N
General description
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
Other Notes
This compound is commonly found in plants of the genus: cannabis
Legal Information
phyproof is a registered trademark of PhytoLab GmbH & Co. KG
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Repr. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service