Skip to Content
Merck
All Photos(1)

Key Documents

VIV105015

SynVivo SynTumour 3D cancer model chip, microvascular network configuration, 8 um pillar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
60104506
NACRES:
NB.35

packaging

case of 1 ea

manufacturer/tradename

SynVivo 105015

storage temp.

room temp

General description

SynTumor cancer model chip- Microvascular design-Barrier type: Pillars, Distance between pillars: 50 μm, Pillar Diameter: 10 μm, Barrier height: 8 μm, Travel/barrier width: 50 μm, Device depth: 100 μm. SynTumor is a 3D tissue model for real-time visualization and quantitative assessment of cell-cell and cell-drug interactions in a physiologically and morphologically realistic tumor microenvironment.

Linkage

LInk to Publications
application text

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service