Skip to Content
Merck
All Photos(1)

Documents

PZ0338

Sigma-Aldrich

Toceranib

≥98% (HPLC)

Synonym(s):

5-[(5Z)-(5-Fluoro-2-oxo-1,2-dihydro-3H-indol-3-ylidene)methyl]-2,4-dimethyl-N-[2-(pyrrolidin-1-yl)ethyl]-1H-pyrrole-3-carboxamide, PHA-291639, SU-11654, SU11654

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H25FN4O2
CAS Number:
Molecular Weight:
396.46
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow to orange

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

room temp

InChI

1S/C22H25FN4O2/c1-13-19(12-17-16-11-15(23)5-6-18(16)26-21(17)28)25-14(2)20(13)22(29)24-7-10-27-8-3-4-9-27/h5-6,11-12,25H,3-4,7-10H2,1-2H3,(H,24,29)(H,26,28)/b17-12-

InChI key

SRSGVKWWVXWSJT-ATVHPVEESA-N

Biochem/physiol Actions

Toceranib (SU11654) is a protein tyrosine kinase inhibitor that selectively targets stem cell factor receptor c-kit, including all forms of mutant Kit, as well as PDGFR and VEGFR. Toceranib exhibited Ki values of 5 nM for PDGFR and 6 nM for VEGFR. Toceranib phosphate is approved for use in mast cell cancer in dogs.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinle Tang et al.
The Biochemical journal, 477(17), 3387-3399 (2020-08-25)
The pseudokinase (PK) RNase L is a functional ribonuclease and plays important roles in human innate immunity. The ribonuclease activity of RNase L can be regulated by the kinase inhibitor sunitinib. The combined use of oncolytic virus and sunitinib has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service