Skip to Content
Merck
All Photos(1)

Documents

C-4 II

88102701, human cervix, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

C-4 II, 88102701

biological source

human cervix

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian cervical carcinoma

Cell Line Description

From a cervical carcinoma of a 41 year old Caucasian female. Cells form spreading predominantly monolayered, epithelial colonies with hemicysts which are characteristic of transporting epithelia and respond to crowding by separation and shedding. These cells have retained properties of basal cell differentiation in long term culture and exhibit G6PD type B. C-4 II cells have been reported to contain human papilloma virus 18 (HPV-18) DNA sequences and to express HPV-18 RNA.

Application

Differentiation studies

Culture Medium

Waymouth′s MB 752/1 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:10 i.e. seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service