Skip to Content
Merck
All Photos(3)

Key Documents

STS0006

Sigma-Aldrich

ECOSURF EH-9

greener alternative

Non-ionic surfactant

Synonym(s):

2-Ethyl hexanol EoPo copolymer (9 EO), Ethyl hexanol ethoxylated propoxylated, Ethylene oxide-propylene oxide copolymer mono(2-ethylhexyl) ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12161902
NACRES:
NA.28
Pricing and availability is not currently available.

grade

laboratory grade

Quality Level

description

non-ionic

form

liquid

greener alternative product characteristics

Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

CMC

1066 ppm (25°C)

viscosity

49.398 cSt(40 °C)

transition temp

pour point 16 °C
cloud point 61 °C (10 wt% actives aq solution)

solubility

water: freely soluble (visual)

density

1.0069 g/mL at 20 °C

HLB

12.5

greener alternative category

General description

This product is an alcohol ethoxylate
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a biodegradable surfactant and is aligned with the 10th principle of Green Chemistry "Design for Degradation".

Features and Benefits

  • Superior wetting
  • Excellent oily soil removal
  • Rapid dissolution & good rinseability
  • Low odor
  • No gel range
  • Outstanding formulation & handling properties
  • Readily biodegradable
  • Aquatic toxicity EC50 > 10 mg/L

Legal Information

ECOSURF is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service