Skip to Content
Merck
All Photos(1)

Key Documents

CDRC00201

Milli-Q

Reverse Osmosis Cartridge

Produces RO purified water at a flow rate of 5 L/hr, For use with Milli-Q® Integral, RiOs Essential, and Elix® Advantage systems

Synonym(s):

Reverse Osmosis Membrane - 5 L/h

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104214
NACRES:
JA.13

packaging

pkg of 1 unit

parameter

5 L/hr flow rate

H × D

28 cm (11.7 in.) × 4.5 cm (1.8 in.)

compatibility

for use with Elix® Advantage
for use with Milli-Q® Integral
for use with RiOs

Looking for similar products? Visit Product Comparison Guide

General description

The Reverse Osmosis membrane removes 95%-99% of contaminants including ions, particulates, bacteria, and organic molecules (MW >200kDa).
For use with Milli-Q® Integral and RiOs Essential, and Elix® Advantage water purification systems.

Application

RO water is used in laboratories, mainly for applications such as general rinsing, feedwater to washing machines and autoclaves, or as a feed to pure and ultrapure water purification systems.

Features and Benefits

Genuine consumable for replacing the Reverse Osmosis Membrane - 5 L/h - CDRC00201.

Legal Information

ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
RIOS is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service