539571
Src Inhibitor, PP1
InSolution, ≥90%
Synonym(s):
InSolution Src Inhibitor, PP1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C16H19N5
Molecular Weight:
281.36
UNSPSC Code:
12352200
NACRES:
NA.77
Recommended Products
Quality Level
Assay
≥90% (HPLC)
form
liquid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light
shipped in
ambient
storage temp.
−20°C
General description
A cell-permeable pyrazolopyrimidine compound that is shown to inhibit Src family tyrosine kinases Lck, Fyn, Hck, and Src (IC50 = 5, 6, 20, and 170 nM, respectively), as well as non-Src family kinases such as CSK, RIP2, and CK1δ (IC50 = 640 nM, 26 nM, 170 nM, respectively), in in vitro kinase assays. At 1 µM, in vitro kinase activity is reduced by more than 57% for p38α, p38β, CK1δ, CSK, RIP2, and GAK, while exhibiting much lower activity against more than 60 other kinases. Kinetic studies employing Src and [poly Glu-Tyr 4:1] indicate that the inhibition is mixed competitive with respect to the substrate and non-competitive with respect to ATP. PP1 also demonstrates anti-growth properties in several B lymphoma cultures of human and murine origins (IC50 = 2.5 to 8.0 µM) and pediatric brain tumor cells (IC50 = 1.1 to 9.7 µM).
A cell-permeable pyrazolopyrimidine compound that is shown to inhibit Src family tyrosine kinases Lck, Fyn, Hck, and Src (IC50 = 5, 6, 20, and 170 nM, respectively), as well as non-Src family kinases such as CSK, RIP2, and CK1δ (IC50 = 640 nM, 26 nM, 170 nM, respectively), in in vitro kinase assays. At 1 µM, in vitro kinase activity is reduced by more than 57% for p38α, p38β, CK1δ, CSK, RIP2, and GAK, while exhibiting much lower activity against more than 60 other kinases. Kinetic studies employing Src and [poly Glu-Tyr 4:1] indicate that the inhibition is mixed competitive with respect to the substrate and non-competitive with respect to ATP. PP1 also demonstrates anti-growth properties in several B lymphoma cultures of human and murine origins (IC50 = 2.5 to 8.0 µM) and pediatric brain tumor cells (IC50 = 1.1 to 9.7 µM). 1 mg of Src Inhibitor, PP1 (Cat. No. 567809) is also available.
Packaging
Packaged under inert gas
Warning
Toxicity: Irritant (B)
Physical form
A 50 mM (5 mg/355 µl) solution of Src Inhibitor, PP1 (Cat. No. 567809) in DMSO.
Reconstitution
Following initial thaw, aliquot and freeze (-20°C).
Other Notes
Ke, J., et.al. 2009. Mol. Cancer8, 132.
Sikkema, A.H., et al. 2009. Cancer Res.69, 5987.
Bain, J., et al. 2007. Biochem. J.408, 297.
Bain, J., et al. 2003. Biochem. J.371, 199.
Karni, R., et al. 2003. FEBS Lett.537, 47.
Hanke, J.H., et al. 1996. J. Biol. Chem.271, 695.
Sikkema, A.H., et al. 2009. Cancer Res.69, 5987.
Bain, J., et al. 2007. Biochem. J.408, 297.
Bain, J., et al. 2003. Biochem. J.371, 199.
Karni, R., et al. 2003. FEBS Lett.537, 47.
Hanke, J.H., et al. 1996. J. Biol. Chem.271, 695.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
188.6 °F - closed cup - (refers to pure substance)
Flash Point(C)
87 °C - closed cup - (refers to pure substance)
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service