Skip to Content
Merck
All Photos(4)

Key Documents

640010P

Avanti

DMABA-d10 NHS Ester

Avanti Research - A Croda Brand

Synonym(s):

4-(dimethylamino)benzoic acid N-hydroxysuccinimide ester-d10

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H4D10N2O4
CAS Number:
Molecular Weight:
272.32
UNSPSC Code:
12352211

description

4-[di(methyl-d3)amino]-2,5-dioxo-1-pyrrolidinyl ester benzoic-2,3,5,6-d4 acid

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (640010P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(ON(C(CC1)=O)C1=O)C2=C([2H])C([2H])=C(N(C([2H])([2H])[2H])C([2H])([2H])[2H])C([2H])=C2[2H]

General description

The DMABA NHS ester reagents were developed by Dr. Robert Murphy′s laboratory at the University of Colorado in order to create PE derivatives where all subclasses and potentially oxidized products could be universally detected using a common precursor ion in the positive ion mode by Mass Spectrometry.

Biochem/physiol Actions

DMABA-d10 NHS Ester, deuterium enriched 10-(dimethylamino) benzoic acid (DMABA) N-hydroxysuccinimide (NHS) ester reagent (d10) is used for derivatizing the primary amine group of PE lipids for differential labeling. It is useful in lipid research. Structurally it has 6 hydrogen atoms deuterated in the 2 methyl groups and 4 hydrogen atoms deuterated in the aromatic ring. d10-DMABA NHS enables detection of phosphatidylethanolamine subclasses in biological samples by tandem mass spectrometry.

Packaging

5 mL Amber Glass Screw Cap Vial (640010P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gilbert O Fruhwirth et al.
Biochimica et biophysica acta, 1772(7), 718-736 (2007-06-16)
Oxidized lipids are generated from (poly)unsaturated diacyl- and alk(en)ylacyl glycerophospholipids under conditions of oxidative stress. The great variety of reaction products is defined by the degree of modification, hydrophobicity, chemical reactivity, physical properties and biological activity. The biological activities of
Karin A Zemski Berry et al.
Analytical chemistry, 81(16), 6633-6640 (2010-03-27)
A set of four (D(0), D(4), D(6), and D(10)) deuterium enriched 4-(dimethylamino)benzoic acid (DMABA) N-hydroxysuccinimide (NHS) ester reagents was developed that react with the primary amine group of glycerophosphoethanolamine (PE) lipids to create derivatives where all subclasses of DMABA labeled
Marko Kosicek et al.
International journal of molecular sciences, 14(1), 1310-1322 (2013-01-12)
Brain is one of the richest organs in lipid content. Phospholipids (glycerophospholipids and sphingolipids) are important building blocks of cell membranes, which provide an optimal environment for protein interactions, trafficking and function. Because of that, alterations in their cellular levels
Yukiko Ueda et al.
Nutrition research (New York, N.Y.), 28(4), 239-244 (2008-12-17)
Inflammatory bowel disease (IBD), including ulcerative colitis (UC) and Crohn disease (CD), is a disorder characterized by diffuse inflammation of the gastrointestinal tract. The immune response and inflammation are mediated by polyunsaturated fatty acids and influenced by dietary fats and
Marcus O W Grimm et al.
International journal of molecular sciences, 14(3), 5879-5898 (2013-03-15)
Alzheimer's disease (AD) is characterized by extracellular accumulation of amyloid-β peptide (Aβ), generated by proteolytic processing of the amyloid precursor protein (APP) by β- and γ-secretase. Aβ generation is inhibited when the initial ectodomain shedding is caused by α-secretase, cleaving

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service