Key Documents
Z682217
Ace receiving flask adapter
joint: ST/NS 29/42, fits Büchi® models R-200/R-205
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
Joint
joint: ST/NS29/42
manufacturer/tradename
Büchi equivalent, 40617
Related Categories
General description
The receiving flask adapter fits condenser assemblies "C", "CR", "A", and "V" for Büchi® evaporator models R-200/205. Adapter clip: not included.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG