NORTCM200
Bruker tapered PTFE caps for Match™ tubes
for NORS205001
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
SB.32
feature
for NORS205001
packaging
pack of 5 ea
manufacturer/tradename
Norell TCM200
Related Categories
General description
Bruker Match™ tube caps are specially machined from PTFE fluoropolymer, are color coded by tube size, and correspond to the color of the tube clamp in the Match™ Insert Assembly, This enables quick identification of matching components. The tube cap incorporates a slightly tapered inner diameter. Initially, the cap aligns and pushes easily onto the tube, it then becomes progressively tighter as the tube reaches full depth within the cap, creating a positive seal to preserve volatile sample solutions throughout short or long term storage.
Legal Information
Match is a trademark of Bruker Biospin Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)