BMSE3000E
Benchmark Accuris™ UV Transilluminator
AC/DC input 230 V, EuroPlug
Synonym(s):
ethidium bromide, gel imaging, gel casting, transillumination
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
viewing area W 16 cm L 19 cm
manufacturer/tradename
Benchmark Scientific E3000-E
spectral range measuring range
302 nm
Looking for similar products? Visit Product Comparison Guide
General description
The Accuris UV Transilluminator is compact in design and offers dual intensity settings (50% and 100%). Optimal for the excitation of Ethidium Bromide, the 302n light source provides even illumination of the viewing surface. Includes hinged UV safety cover.
Application
Electrophoresis
Legal Information
Accuris is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service