Skip to Content
Merck
All Photos(1)

Key Documents

933058

Sigma-Aldrich

EMS SMD Precision Tweezer Kit

SS

Synonym(s):

grippers kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NB.35

General description

EMS techware

Application

EMS SMD Precision Tweezer Kit, SS are small hand tools used for grasping objects too small to be easily handled with the human fingers. We offer Kit of 5 SMD tweezers - Anti-Acid/Anti-Mag stainless steel (SS) with top quality hard/foam wallet.
Includes styles: SM103, SM107, SM108, SM111, SM115.
To improve your productivity and allow you to view more than one sample at a time

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service