Skip to Content
Merck
All Photos(1)

Key Documents

904252

Sigma-Aldrich

Sodium taurocholate-2,2,4,4-d4 solution

100 μg/mL in methanol, ≥98 atom % D, ≥98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26D4H40NO7SNa
Molecular Weight:
541.71
UNSPSC Code:
12352106
NACRES:
NA.24

isotopic purity

≥98 atom % D

Quality Level

Assay

≥98% (CP)

form

liquid

concentration

100 μg/mL in methanol

technique(s)

mass spectrometry (MS): suitable

mass shift

M+4

storage temp.

−20°C

SMILES string

O=C(NCCS(=O)(O[Na])=O)CC[C@H]([C@@H](CC1)[C@]([C@@H](O)C2)(C)C1C([C@H](O)C3)C2[C@]4(C)[C@@]3([H])C([2H])([2H])[C@H](O)C([2H])([2H])C4)C

Related Categories

Packaging

1 mL in ampule
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service