About This Item
Assay
96%
reaction suitability
core: chromium
reagent type: catalyst
mp
54-59 °C (lit.)
SMILES string
[O]#C[Cr](C#[O])C#[O].CCOC(=O)c1ccccc1
InChI
1S/C9H10O2.3CO.Cr/c1-2-11-9(10)8-6-4-3-5-7-8;3*1-2;/h3-7H,2H2,1H3;;;;
InChI key
SWSAJUVURPSGIG-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service