All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H10N2
CAS Number:
Molecular Weight:
110.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
refractive index
n20/D 1.48 (lit.)
bp
138-140 °C (lit.)
density
0.912 g/mL at 25 °C (lit.)
functional group
amine
SMILES string
CN(C)n1cccc1
InChI
1S/C6H10N2/c1-7(2)8-5-3-4-6-8/h3-6H,1-2H3
InChI key
ZOORJGVSPXCZCJ-UHFFFAOYSA-N
General description
1-(Dimethylamino)pyrrole is a pyrrole monomer and can be polymerized electrochemically or by using simple chemical polymerization methods to form electroactive polymers.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
98.6 °F - closed cup
Flash Point(C)
37 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Thiophene and Pyrrole Monomers.Modern Polymerization Techniques: 28.
Hydroxymethyl EDOT.
Material Matters, 5(1) (2010)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service