Skip to Content
Merck
All Photos(1)

Key Documents

228818

Sigma-Aldrich

Diethyl propylmalonate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH(COOC2H5)2
CAS Number:
Molecular Weight:
202.25
Beilstein:
510157
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.418 (lit.)

bp

221-222 °C (lit.)

density

0.987 g/mL at 25 °C (lit.)

SMILES string

CCCC(C(=O)OCC)C(=O)OCC

InChI

1S/C10H18O4/c1-4-7-8(9(11)13-5-2)10(12)14-6-3/h8H,4-7H2,1-3H3

InChI key

GRRSDGHTSMJICM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diethyl propylmalonate was used in the synthesis of propylacrylic acid (PAA).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamed E H El-Sayed et al.
Journal of controlled release : official journal of the Controlled Release Society, 104(2), 417-427 (2005-06-29)
Limited cytoplasmic delivery of enzyme-susceptible drugs remains a significant challenge facing the development of protein and nucleic acid therapies that act in intracellular compartments. "Smart" pH-responsive, membrane-destabilizing polymers present a new approach to shuttling therapeutic molecules past the endosomal membrane
Kyle M Hocking et al.
Tissue engineering. Part A, 25(5-6), 416-426 (2018-08-23)
Subarachnoid hemorrhage (SAH) is associated with vasospasm that is refractory to traditional vasodilators, and inhibition of vasospasm after SAH remains a large unmet clinical need. SAH causes changes in the phosphorylation state of the small heat shock proteins (HSPs), HSP20
Feng Qiu et al.
Biomaterials, 182, 82-91 (2018-08-15)
Cancer vaccines targeting patient-specific tumor neoantigens have recently emerged as a promising component of the rapidly expanding immunotherapeutic armamentarium. However, neoantigenic peptides typically elicit weak CD8+ T cell responses, and so there is a need for universally applicable vaccine delivery
Brian C Evans et al.
ACS nano, 9(6), 5893-5907 (2015-05-26)
A platform technology has been developed and tested for delivery of intracellular-acting peptides through electrostatically complexed nanoparticles, or nano-polyplexes, formulated from an anionic endosomolytic polymer and cationic therapeutic peptides. This delivery platform has been initially tested and optimized for delivery
Xi Zhan et al.
Pharmaceutical research, 32(7), 2280-2291 (2015-01-17)
A key step of delivering extracellular agents to its intracellular target is to escape from endosomal/lysosomal compartments, while minimizing the release of digestive enzymes that may compromise cellular functions. In this study, we examined the intracellular distribution of both fluorecent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service