Skip to Content
Merck
All Photos(2)

Key Documents

SAB2108778

Sigma-Aldrich

Anti-SRRM2 (C-terminal) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

bHLHe14

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

34 kDa

species reactivity (predicted by homology)

horse, mouse, bovine, canine, human, guinea pig, rabbit, pig

concentration

0.5 mg/mL

technique(s)

western blot: 1 μg/mL

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SRRM2(57396)

General description

SRRM2 is involved in pre-mRNA splicing. It may function at or prior to the first catalytic step of splicing at the catalytic center of the spliceosome and may do so by stabilizing the catalytic center or the position of the RNA substrate. It binds to RNA.

Immunogen

Synthetic peptide directed towards the C-terminal region of Human SRRM2

Sequence

Synthetic peptide located within the following region: DKKEKSATRPSPSPERSSTGPEPPAPTPLLAERHGGSPQPLATTPLSQEP

Physical form

Supplied at 0.5 mg/ml in phosphate-buffered saline, 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soňa Legartová et al.
Cells, 10(2) (2021-02-05)
The essential components of splicing are the splicing factors accumulated in nuclear speckles; thus, we studied how DNA damaging agents and A-type lamin depletion affect the properties of these regions, positive on the SC-35 protein. We observed that inhibitor of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service