OH30241168
Ohaus® MB90 Mositure Analyzer
Model MB90, weighing capacity 90 g, resolution: 0.001 g, AC/DC input 210 - 2310 V, Schuko plug
Synonym(s):
Ohaus® Moisture Analyzer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41111500
NACRES:
NB.13
Recommended Products
material
metal weighing plate
feature
CE compliant
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 30241168
weighing capacity measuring range
90 g, resolution: 0.001 g
weighing pan diam.
Looking for similar products? Visit Product Comparison Guide
General description
Combining Speed and Precision in One Moisture Analyzer
Application
Percent Moisture Determination, Percent Solid Determination, Weighing
Features and Benefits
Colour touchscreen display with intuitive icon-based navigation Fast halogen heating technology provides uniform drying for precise and repeatable results Analysis made simple: tare, add your sample, press start and see your results Robust hou
Legal Information
Ohaus is a registered trademark of Ohaus Corp.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service