EP0030505107
Eppendorf® Deepwell plates, standard, 96 wells
white plate, conical bottom colorless wells, capacity 500 μL, pkg of 120 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122107
NACRES:
NB.15
material:
conical bottom colorless wells
polypropylene
white plate
polypropylene
white plate
size:
96 wells
sterility:
sterile; autoclaved
Recommended Products
material
conical bottom colorless wells
polypropylene
white plate
sterility
sterile; autoclaved
packaging
pkg of 120 ea
manufacturer/tradename
Eppendorf® 0030505107
availability
available only in (India)
technique(s)
PCR: suitable
capacity
500 μL
size
96 wells
working volume
500 μL
Looking for similar products? Visit Product Comparison Guide
General description
For greater reliability, stability and efficiency, choose Eppendorf Deepwell 96 well Plates. Available with white, yellow, green or blue frames, these deepwell plates offer high performance for all applications - both manual and automated. Thanks to OptiTrack matrix, a high-contrast alphanumeric label laser-engraved into the frame, you can find your sample up to 30% faster and reduce pipetting errors. For every application, from sample storage at 86 C to DNA denaturation at 100 C, rely on Eppendorf Deepwell Plates and set new standards in your laboratory.
Application
- Sample storage and preparation Bacteria and yeast cultivation
- High-throughput DNA and RNA isolation
- Storage of cDNA or genomic libraries
- Storage of siRNA or oligonucleotide libraries
- Protein analysis Storage of valuable protein and DNA samples in Eppendorf LoBind Plates
- High-throughput plasmid purification Creation of dilution series
Features and Benefits
- OptiTrack matrix: up to 30 % faster sample detection and fewer pipetting errors thanks to high-contrast alphanumeric labeling
- RecoverMax well design: Optimized well geometry for maximum sample recovery and excellent mixing properties
- g-Safe : exceptional centrifugation resistance up to 6,000 × g for faster protocols and improved sample quality
- Raised corrugated edges and smooth surface for reliable sealing, even during heat sealing
- Easily and reliably stackable
- Minimal residual volume and high homogeneity from well to well
- High-quality polypropylene for high resistance to chemicals, mechanical stress and temperature extremes
- Manufactured without mold release agents, plasticizers and biocides – substances that can negatively affect bioassays
- Autoclavable (121 C, 20 min)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service