Skip to Content
Merck
All Photos(1)

Key Documents

AXYTE1004B

Pipette tips for Eppendorf® and Socorex pipettors

size 1000 μL, blue, pkg of 10x500tips/cs

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
manufacturer/tradename:
Axygen Inc TE-1004-B
Corning
size:
1000 μL
sterility:
non-sterile
packaging:
pkg of 10x500tips/cs

sterility

non-sterile

manufacturer/tradename

Axygen Inc TE-1004-B
Corning

packaging

pkg of 10x500tips/cs

size

1000 μL

color

blue

Looking for similar products? Visit Product Comparison Guide

Other Notes

Browse all pipette tips

Legal Information

Corning is a registered trademark of Corning, Inc.
Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service